BLAUBRAND® Volumetric Flasks, Class A, Blue Markings
Supplier: Brand
37256 37264 37265 37266 37288 37289 37294 37259 37260 37267 37258 37262 37263 37257 37261 37268
612-2738EA 43.3 EUR
612-2738 827-6453 612-2742 612-2743 612-2745 612-2746 612-2747 827-6448 827-6449 612-2744 827-6447 612-2741 827-6452 612-2739 612-2740 612-3654
BLAUBRAND® Volumetric Flasks, Class A, Blue Markings
Flasks Volumetric Flasks
Borosilicate glass 3.3, with glass stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
*Wide neck.
Certifikace: DIN EN ISO 1042
Informace pro objednávání: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.