Vacuum tweezers, aspirette®
Supplier: HIRSCHMANN
9901500 9901600 9901601 9901700 9901605
232-2420EA 255 EUR
232-2420 HIRS9901600 HIRS9901601 HIRS9901700 HIRS9901605
Vacuum tweezers, aspirette®
Forceps
Powered by rechargeable battery for picking up and placing flat objects up to a max. unit weight of 70 g.
  • One-button operation, ergonomic shape
  • Standard and antistatic models available
  • Rechargeable battery can be charged ∼1000 times
Informace pro objednávání: The package includes a mains charger and a suction unit set consisting of three suction nozzles with 3,2, 6 and 10 mm Ø suction cups made of silicone.