Funnels with long stem
Supplier: SciLabware
2145/06M 2145/03M 2145/10M
221-0024EA 37 EUR
221-0024 221-0023 221-0025
Funnels with long stem
Funnels Filtration Funnels Filter Funnels
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls. Cone angle 60°.