Funnels with short stem, Nalgene®
Supplier: Thermo Fisher Scientific
4260-0040 4260-0060 4260-0100
221-0096EA 42.6 EUR
221-0096 221-0097 221-0098
Funnels with short stem, Nalgene®
Funnels Filtration Funnels Filter Funnels
LDPE, transparent.
  • Good chemical resistance
  • Include external ribs for better air evacuation and a suspension eye
  • Ideal for filtration with partial vacuum
With smooth internal walls. Very sturdy construction.