Büchner funnels
Supplier: KARTELL
437 438 439 440 441 442 443 445
519-2361EA 25.6 EUR
519-2361 519-2362 519-2363 511-0008 519-2364 519-2365 519-2366 519-2367
Büchner funnels
Funnels Filtration Funnels Büchner Funnels
PP, white.
  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning
For filter paper or membrane filtration discs.