Filter Funnels
Supplier: ROBU GLASFILTER
21501 21502 21503 21504 21505 21751 21752 21753 21754 21755 21802 21121 21122 21123 21124 21125 21551 21552 21553 21554 21555 21111 21112 21113 21114 21115 21401 21402 21403 21404 21400 21120 21500 21550 21750 21800 21801 21803 21804 21110 21254 21253 21252 21251 21250 21255 2112F 2111F 2125F 2150F 2155F 2175F
511-1301EA 37.8 EUR
511-1301 511-1302 511-1303 511-1304 511-1305 511-1311 511-1312 511-1313 511-1314 511-1315 511-1316 511-1321 511-1322 511-1323 511-1324 511-1325 511-1331 511-1332 511-1333 511-1334 511-1335 511-1341 511-1342 511-1343 511-1344 511-1345 511-1351 511-1352 511-1353 511-1354 511-0370 511-0363 511-0371 511-0372 511-0373 511-0374 511-0375 511-0376 511-0377 511-0362 511-0368 511-0367 511-0366 511-0365 511-0364 511-0369 ROBU2112F ROBU2111F ROBU2125F ROBU2150F ROBU2155F ROBU2175F
Filter Funnels
Funnels Filtration Funnels Büchner Funnels
ROBU® Filter Funnel with VitraPOR® Filter is designed for high-performance filtration. Equipped with a VitraPOR® filter, it offers exceptional chemical resistance and precision, making it ideal for demanding laboratory and industrial filtration processes. The funnel is durable, reusable, and easy to clean.
  • VitraPOR® filter for superior performance
  • High chemical resistance
  • Precise filtration
  • Durable and reusable
  • Ideal for laboratory and industrial use
  • Easy to clean
  • Efficient filtration for demanding applications
  • Robust construction for long-lasting use
The ROBU Filter Funnel with VitraPOR filter is engineered for high-efficiency filtration in both laboratory and industrial settings. Featuring the VitraPOR filter, known for its exceptional chemical resistance and precision, this funnel delivers reliable performance in demanding filtration processes. Its robust construction ensures durability, while the funnel is easy to clean and can be reused, providing an environmentally friendly and cost-effective solution. Designed for use with various filter papers or membranes, it ensures optimal filtration, making it ideal for applications requiring precise and efficient result
Certifikace: In compliance with ISO 4796-80 and ASTM E128-99 standards.