Filter Funnels, Conical
Supplier: ROBU GLASFILTER
22251 22252 22253 22254 22255 22261 22262 22263 22265 22264
511-3301EA 23.3 EUR
511-3301 511-3302 511-3303 511-3304 511-0378 511-0379 511-0380 511-0381 511-0383 511-0382
Filter Funnels, Conical
Funnels Filtration Funnels Filter Funnels
ROBU® filter funnel conical with intergrated VitraPOR® filter is a classic laboratory tool designed for reliable filtration. Made from high-quality borosilicate glass, it ensures chemical resistance and durability across a wide range of applications.
  • Conical design
  • Precise pour control
  • Borosilicate glass
  • Chemical resistance
  • Heat resistance
  • Reusable
  • Suitable for gravity filtration
  • Compatible with standard filter papers
The ROBU funnel with integrated VitraPor filter, conical, is an essential component for laboratory filtration processes. Its tapered design promotes smooth and controlled fluid flow, making it suitable for both qualitative and quantitative filtration. Manufactured from durable borosilicate glass, the funnel offers excellent resistance to thermal shock and aggressive chemicals.
This reusable funnel is compatible with standard filter papers and ideal for a variety of laboratory settings including research, quality control, and educational environments.
Certifikace:
In compliance with ISO 4796-80 and ASTM E128-99 standards.