Membrane filter funnels, magnetic
Supplier: Cytiva (Formerly Pall Lab)
4238 4241 4247 4242-N
516-7590EA 407 EUR
516-7590 516-7593 516-7597 516-7594
Membrane filter funnels, magnetic
Funnels Membrane Filter Funnels
Polyphenylsulfone, autoclavable.
  • Leakproof magnetic seal allows one-handed operation
  • Compatible with anti-foaming agents and many other solvents
For membrane filtration techniques for water testing.
Effective filtration area: 150 and 300 ml: 9,6 cm²; 500 ml: 13,1 cm²
Overall Ø×H: 150 ml: 76×178 mm; 300 ml: 76×229 mm; 500 ml: 89×196 mm