Vacuum membrane filter holder
Supplier: Cytiva (Formerly Pall Lab)
4013 4011-N 4012-N
516-8921EA 797 EUR
516-8921 514-4000 516-7563
Vacuum membrane filter holder
Funnels Membrane Filter Funnels
Borosilicate glass, aluminium clamp.
Ideal for vacuum filtration of liquids and degassing of HPLC solvents and mobile phase. With graduation.
Filtration area: 9,6 cm².
* with stopper support base, without flask.
Delivery information: Filtration funnel with No. 8 stopper support base (514-4000); filtration unit with side arm support assembly NS 40/35, with flask (516-8921).