Cleanroom gloves, latex, sterile, BioClean Extra™
Supplier: Nitritex
BLAS-L BLAS-M BLAS-S BLAS-XL BLAS-L-P1
112-3078EA 730 EUR
112-3078 112-3079 112-3080 112-3081 NITRBLAS-L-P1
Cleanroom gloves, latex, sterile, BioClean Extra™
Gloves
Ambidextrous cleanroom gloves with beaded cuffs. Suitable for double donning.
  • Sterile, powder-free, textured finish
  • Extended length provides protection up to the elbow
  • Protein level less than 50 µg/g
  • Extended leaching in pure deionised water ensures extremely low levels of chlorides and extractable metals
  • Each size carries a unique barcode on the carton (ITF-14), which allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
Length: 400 mm
Thickness: 0,10 mm (cuff); 0,13 mm (palm); 0,16 mm (fingers)
Colour: Natural
Certifikace: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003, +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Balení: One cuffed pair per inner plastic wallet; one inner wallet per EasyTear™ PE pouch; 10 pairs per sealed outer PE bag; 20 outer bags per carton (200 pairs).