Cleanroom gloves, nitrile, Chemsoft CE™
Supplier: North by Honeywell
Chemsoft CE™
112-2550EA 95.1 EUR
112-2550 112-2548 112-2549 112-2546 112-2547
Cleanroom gloves, nitrile, Chemsoft CE™
Gloves
These gloves have anti-static properties and are therefore ideal for the fine assembly of parts in cleanrooms, pharmaceutical areas and paints shop.
  • Non sterile, powder-free
  • Anti-static and state-of-the-art cleaning process reduces particulate contamination
  • Ambidextrous and textured fingertips provide a good grip
  • Rolled cuff prevents tearing while donning
Thickness: 0,10 mm
Colour: White
Balení: Double bagged to ensure cleanliness.
Certifikace: EN ISO 374-1 Type B (JKT); Cat. III; AQL 1,5
Balení: Double bagged to ensure cleanliness.