Chemical and heat resistant gloves, AlphaTec® 19-024 / 19-026
Supplier: Ansell
19-026/8 19-024/8 19-024/10 19-026/10
112-2307EA 111 EUR
112-2307 112-2396 112-2397 112-2398
Chemical and heat resistant gloves, AlphaTec® 19-024 / 19-026
Gloves
These gloves combine the flexibility and chemical resistance of neoprene with the comfort and heat resistance of a soft thermal liner. They are designed to protect users in applications where thermal and chemical risks are present.
  • Neoprene coating protects against a wide range of chemicals
  • Excellent mechanical and chemical properties (protects against greases and oils)
  • Comfortable insulating double liner provides protection from intermittent contact with hot surfaces up to 180 °C
Length: 460 or 660 mm
Colour: Black
EN 374 (AKL), EN 388 (3222), EN 407 (32XX4X), EN 511 (121), Cat. III; AQL 1.0
Certifikace: EN ISO 374-1:2016 Type A (AKLMPT), EN ISO 374-5:2016, EN 388:2016 (3122B), EN 407 (32XX4X), EN 511 (121), Cat. III; AQL 1.0
Upozornení: Not for use in or around open flame.