Heat resistant gloves
Supplier: GANTERIE CO KG
14120 141220
112-2995EA 76.7 EUR
112-2995 112-2996
Heat resistant gloves
Gloves
Manufactured from Nomex® fabric, using non-stop seamless process. Small loop terry towel fabric.
  • Good mechanical resistance to abrasion and cuts
  • Ambidextrous shape doubles glove life
  • Excellent resistance to flames and small molten metal splashes
For handling sharp, hot components up to 250 °C. Ideal for welding or where there is a risk of hot projectiles.
Colour: Grey
EN 388, EN 407
Certifikace: EN 388, EN 407