Heat and cold resistant gloves
Supplier: GANTERIE CO KG
1100 1200
112-1835EA 129 EUR
112-1835 112-1836
Heat and cold resistant gloves
Gloves
These knitted gloves made from carbon silicon fibres protect against heat and cold.
  • Cold resistant down to −80 °C
  • Heat resistant up to 95 °C
  • Unsuitable for use with liquid nitrogen
Ideal for handling dry ice or working in a freezer.
EN 407
Certifikace: EN 407