Dry box gloves, neoprene
Supplier: PIERCAN
97800C96/10 10750C84/10 10500C76/10 10500C86/10 10750C74/10 14800C76/10 14800C86/10 14800C96/10 14800C84/10 14800C94/10 10500C96/10 18800C96/10 18800C94/10 18800C86/10 18800C84/10 18800C106/10 18800C104/10 10750C94/10 10750C106/10 10750C104/10 13800C96/10 13800C106/10 10750C76/10 10750C86/10 10750C96/10 13800C76/10 13800C86/10 10500C104/10 10500C106/10 10500C74/10 10500C84/10 10500C94/10 13800C104/10 13800C74/10 13800C84/10 13800C94/10 14500C6E66 14500C7E46 14500C7E66 14500C8E46 14500C8E66 14500C9E46 14500C9E66 14800C6E46 18800C94/6 10750C8E46 10750C9E46
PIEC97800C96/10EA 3970 EUR
PIEC97800C96/10 PIEC10750C84/10 PIEC10500C76/10 PIEC10500C86/10 PIEC10750C74/10 PIEC14800C76/10 PIEC14800C86/10 PIEC14800C96/10 PIEC14800C84/10 PIEC14800C94/10 PIEC10500C96/10 PIEC18800C96/10 PIEC18800C94/10 PIEC18800C86/10 PIEC18800C84/10 PIEC18800C106/10 PIEC18800C104/10 PIEC10750C94/10 PIEC10750C106/10 PIEC10750C104/10 PIEC13800C96/10 PIEC13800C106/10 PIEC10750C76/10 PIEC10750C86/10 PIEC10750C96/10 PIEC13800C76/10 PIEC13800C86/10 PIEC10500C104/10 PIEC10500C106/10 PIEC10500C74/10 PIEC10500C84/10 PIEC10500C94/10 PIEC13800C104/10 PIEC13800C74/10 PIEC13800C84/10 PIEC13800C94/10 PIEC14500C6E66 PIEC14500C7E46 PIEC14500C7E66 PIEC14500C8E46 PIEC14500C8E66 PIEC14500C9E46 PIEC14500C9E66 PIEC14800C6E46 PIEC18800C94/6 PIEC10750C8E46 PIEC10750C9E46
Dry box gloves, neoprene
Gloves
Ambidextrous gauntlets manufactured from soft black polychloroprene rubber that is self-extinguishing thus providing a good flame resistance for the gauntlet. It is the ideal elastomer providing the best compromise to the conjugate action of several potentially harmful agents, for example oil and ozone. Available for use with various diameter ports.
  • High tensile strength and flexibility
  • Good resistance to the effects of ozone ageing, oxygen and weathering
Colour: Black
0,4 mm thickness: EN 388 (X100), EN 374-1, EN 421; Cat. III
0,6 mm thickness: EN 388 (X100), EN 374-1 (ACK), EN 421; Cat. III
0,8 mm thickness: EN 388 (X111), EN 374-1 (ACK), EN 421; Cat. III
Certifikace: 0,4 mm thickness: EN ISO 374-1 Type B (ANP), EN ISO 374-5 VIRUS, EN 421; Cat. III
0,6 mm thickness: EN 388:2016 (X1XXX), EN ISO 374-1 Type B (ANP), EN ISO 374-5 VIRUS, EN 421; Cat. III
0,8 mm thickness: EN 388:2016 (X111X), EN ISO 374-1 Type B (ANP), EN ISO 374-5 VIRUS, EN 421; Cat. III