Goggles, Attack
Supplier: Bollé Safety
Attack
111-0313EA 18.3 EUR
111-0313
Goggles, Attack
Goggles
These goggles offer a panoramic vision and anti-scratch coating on both sides. They provide protection against impact, chemical splash, dust and liquid aerosols.
  • Cushioned frame provides maximum comfort and protection
  • Fitted with a wide, adjustable headband
  • Ideal for use with prescription spectacles due to the temple gaps
  • Can be used with certain respirators
Weight: 95 g
Certifikace: EN 166, EN 170.