Goggles, V-Maxx
Supplier: HONEYWELL SAFETY
1006193 1006192 1007506 1006194 1006195 1006196 1008111
111-3585EA 16.3 EUR
111-3585 111-2295 111-0736 111-2296 111-2297 111-2298 111-2299
Goggles, V-Maxx
Goggles
Sporty design goggles with elastic strap.
  • 180° field of vision for distortion-free peripheral vision
  • Rotating strap attachments for easier use with hardhats
  • Indirect ventilation complies with splash-resistance test
Can be worn over prescription eyewear and some half mask respirators.
Weight: 93 g.
Certifikace: EN 166, EN 169, EN 170, 1 F, C, 3, 4, 9