Hydrometers for mineral oils
Supplier: GERING
0429WG 0431WG 0428WG 0430WG 0432WG
614-0429EA 121 EUR
614-0429 614-0431 614-0428 614-0430 614-0432
Hydrometers for mineral oils
Hydrometers
Hydrometer for mineral oils with thermometer contained within the stem.
  • Calibration temperature: 15 °C