Staining jars acc. to Hellendahl, WHEATON®
Supplier: DWK Life Sciences
720-0711EA 374 EUR
720-0711
Staining jars acc. to Hellendahl, WHEATON®
Jars Staining Jars
Soda-lime glass.
  • Wide top for convenient transfer
  • For staining slides that are inscribed on one end
These staining jars hold up to eight single standard slides (76×26 mm) positioned vertically or 16 slides back to back.