Drums, conductive, with screw lid
Supplier: CURTEC
7026-63-101
216-5810EA 141 EUR
216-5810
Drums, conductive, with screw lid
Barrels, Kegs, Drums
HDPE conductive black drums, with black screw lids.
  • Surface resistivity ≤ 1,000,000 Ohm
  • Water tight
  • Tamper evident
  • Stackable
  • UN marking (1H2) for hazardous solids
For storage and shipment of explosive powders.