Full face masks, 6000, 3M
Supplier: 3M
6700S 6800M 6900L
113-0244EA 266 EUR
113-0244 111-0206 111-0207
Full face masks, 6000, 3M
Respirators Reusable Respirators Full Face Respirators
Lightweight full face masks for maximum comfort. Incorporating a soft, hypoallergenic material and central exhalation valve.
  • Uses 6000/2000 series bayonet filters
  • A large lens, coated for scratch resistance, provides excellent visibility with a wide field of vision
  • Cool Flow® exhalation valve is designed to make breathing easier and to help reduce heat and moisture build-up in the face piece
  • Low maintenance respirator: Can be disassembled, cleaned and reused many times
  • No replacement parts are necessary except filters and cartridges
Weight: 400 g
EN 136
Weight: 400 g
Certifikace: EN 136
Informace pro objednávání: Supplied without filters, which are sold separately.