Full face masks, BRK 820
Supplier: Scott Safety
111-3060EA 109 EUR
111-3060
Full face masks, BRK 820
Respirators Reusable Respirators Full Face Respirators
This reusable full face mask is for use in industry, agriculture, rescue work and the disposal of hazardous substances. In addition to its use as a sterile filter mask, the mask also functions as a respiration mask for use with lightweight tubing equipment thanks to its standard round thread connection, in accordance with EN 148-1.
  • Visibility is maximised due to the low, space-optimising connectors; viewing pane made from heat-resistant polycarbonate
  • Integrated dampers prevent injury caused by impact
  • With polyester speech diaphragm and five-point head-strap
  • Two exhalation valves ensure low resistance when exhaling; optimal airflow ensures an anti-fog view
  • Choice of skin-friendly, highly durable rubber or silicone, specifically for allergy sufferers
Certifikace: EN 136, EN 148
Upozornení: *One size fits all