Gas, vapour and particle filters for half and full face masks
Supplier: HONEYWELL SAFETY
1785010 1781000 1784000 1784140 1783040
111-3589EA 28.9 EUR
111-3589 111-3587 111-3588 111-0136 111-0253
Gas, vapour and particle filters for half and full face masks
Respirators Reusable Respirators Respirator Filters
Gas filters with bayonet attachment for use with half masks.
  • Easy to replace
  • Low respiratory resistance
A: Organic vapours with boiling points over 65 °C
AX: Organic vapours with boiling points under 65 °C
B: Inorganic gases and vapours, e.g. chlorine, hydrogen cyanide, hydrogen sulphide
E: Acidic gases, e.g. sulphur dioxide, hydrogen chloride
K: Ammonia
Certifikace: EN 14387, EN143