Measuring cylinders, tall form, class B, PMP, Nalgene®
Supplier: Thermo Fisher Scientific
3663-0010 3663-0025 3663-0050 3663-0100 3663-0250 3663-0500 3663-1000 3663-2000 3663-4000
612-4010EA 19.2 EUR
612-4010 612-4011 612-4012 612-4013 612-4014 612-4015 612-4016 612-4017 612-4018
Measuring cylinders, tall form, class B, PMP, Nalgene®
Cylinders Measuring Cylinders
PMP, transparent, tall form, autoclavable, class B.
  • Calibrated to contain (TC, In) at 20 °C, non-wetting - no meniscus to confuse readings
  • Moulded graduations and hexagonal base in blue PP, stable, large spout, very robust
  • Autoclavable, but accuracy will be affected
ASTM Class B E1272 and ISO 6706
Certifikace: ASTM Class B E1272 and ISO 6706