Measuring cylinders, tall form, class B, borosilicate glass
Supplier: HIRSCHMANN
2200154 2200160 2200170 2200175 2200180 2200186 2200190 2200193 2200194 2200197
612-3027EA 14.4 EUR
612-3027 612-3028 612-3029 612-3030 612-3031 612-3032 612-3033 612-3034 612-3035 612-3036
Measuring cylinders, tall form, class B, borosilicate glass
Cylinders Measuring Cylinders
DURAN®, borosilicate glass, tall form, class B.
  • Calibrated to contain 'ln'
  • Hexagonal glass base, with spout
  • Amber stain graduations
* In addition to DIN.
Certifikace: DIN EN ISO 4788