Measuring cylinders, tall form, class A, borosilicate glass
Supplier: HIRSCHMANN
2270193 2270160 2270170 2270175 2270180 2270186 2270190
613-4717EA 62.3 EUR
613-4717 612-3360 612-3361 612-3362 612-3363 612-3364 612-3365
Measuring cylinders, tall form, class A, borosilicate glass
Cylinders Measuring Cylinders
DURAN®, borosilicate glass, tall form, class A.
  • Calibrated to contain 'ln', certificate of conformity (DE-M marking) with dated batch identification available via website: (www.hirschmannlab.de)
  • Hexagonal plastic base and protective collar with spout
  • Short graduation marks, blue graduations
DIN EN ISO 4788
Certifikace: DIN EN ISO 4788