Immunology plates and modules, Nunc Immobilizer™
Supplier: Thermo Fisher Scientific
436014 436006 436007 436013 436008 436015 436016 436017 436018 436020 436022 436023
734-1284EA 577 EUR
734-1284 734-1277 734-1278 734-1283 734-1279 734-1285 734-1286 734-1287 734-1288 734-1290 734-1291 734-1292
Immunology plates and modules, Nunc Immobilizer™
Microplates Immunology Microplates and Strips
PS, non sterile, without lid. Available with a choice of surface chemistries: Amino surface for rapid coupling of peptides and proteins, streptavidin surface for rapid coupling of biotinylated biomolecules such as peptides, antibodies, oligonucleotides or haptens. Also available clear, for colorimetric assays and sample storage, white, for luminescence and fluorescence assays, or black, for fluorescence assays.
  • Ready to use plates and modules requiring no activation or blocking steps
  • Robust and stable surfaces with long shelf life
  • High sensitivity and specificity
  • High specific binding
  • High signal to noise ratio
Total well volume: 400 µl (96-well plates and flat well strips), 350 µl (C-well strips), 120 µl (384-well plates).
Total well volume: 400 µl (96-well plates and flat well strips), 350 µl (C-well strips), 120 µl (384-well plates)