MicroWell™ Plates, 96 well, FluoroNunc™ and LumiNunc™
Supplier: Thermo Fisher Scientific
136101 136102 137101 137103
734-2002EA 264 EUR
734-2002 734-2003 734-2017 734-2018
MicroWell™ Plates, 96 well, FluoroNunc™ and LumiNunc™
Microplates HTS Microplates
PS, tissue culture-treated, sterile, with lid. 96-MicroWell™ plates for fluorescence and luminescence based cell assays. Black plates for fluorescence reading with minimum back-scattered light and background fluorescence. White plates for luminescence with maximum reflection and minimal autoluminescence.
  • Raised well rims reduce risk of cross-contamination
  • Flat (F) bottom wells for optimum optical characteristics
  • Plate edges and lids designed to reduce evaporation
  • Plates fit all standard equipment
  • Nunclon™Δ certified treated surface
Recommended working volume: 200 µl per well
Recommended working volume: 200 µl per well