MicroWell™ Plates, 96 well, FluoroNunc™ and LumiNunc™
Supplier: Thermo Fisher Scientific
236105 236107 236108 237105 237107 237108
732-2696EA 226 EUR
732-2696 732-2697 732-2698 732-2699 732-2700 732-2701
MicroWell™ Plates, 96 well, FluoroNunc™ and LumiNunc™
Microplates HTS Microplates
PS, untreated, pinchbar design. 96-MicroWell™ plates for fluorescence and luminescence based assays. Black plates for fluorescence reading with minimum back-scattered light and background fluorescence. White plates for luminescence with maximum reflection and minimal autoluminescence.
  • Raised well rims reduce risk of cross-contamination
  • Flat (F) bottom wells for optimum optical characteristics
  • Plates fit all standard equipment
  • Compatible with lids for MicroWell™ plates and adhesive sealing tape
Recommended working volume: 200 µl per well
Recommended working volume: 200 µl per well