Microplates, 96- and 384-well, Nunc™
Supplier: Thermo Fisher Scientific
249943 249944 249945 249946 249947 249949 249950 264573 264575 264576 264579 267245 267334 267342 267350 267369 267385 442587
732-2619EA 300 EUR
732-2619 732-2620 732-2621 732-2622 732-2623 732-2624 732-2625 732-2654 732-2656 732-2657 732-2660 732-2661 732-2662 732-2663 732-2664 732-2665 732-2666 735-0016
Microplates, 96- and 384-well, Nunc™
Microplates HTS Microplates
PP, untreated, without lid. These plates are ideal for liquid phase assays, sample preparation, compound storage and chemical synthesis as they are non toxic and resistant to most chemicals, solvents and alcohols. White plates for luminescence applications maximise reflection of signal and minimise autoluminescence. Black plates for fluorescence applications reduce background fluorescence and minimise back-scattered light. Coloured plates are ideal for storage applications.
  • Compatible with robotics and automated systems
  • Choice of conical (V) or round (U) bottom well shape for optimal sample recovery
  • Rounded square wells minimise wicking
  • Shared wall technology of 96-MicroWell™ plate for space saving and improved mixing in wells (except Cat. No. 735-0016)
*Certified free of RNase and DNase.