Cell culture inserts, Nunc™
Supplier: Thermo Fisher Scientific
140620 140640 140627 140629 140642 140644 140652 140654 140656 140660 140663 140668
734-2239EA 442 EUR
734-2239 734-2240 734-2228 734-2229 734-2230 734-2231 734-2232 734-2233 734-2234 734-2235 734-2236 734-2237
Cell culture inserts, Nunc™
Microplates Insert Plates and Inserts
For cultivation of most cell types, without matrix coating. Membranes are thermally welded to the polystyrene upper structure. Polycarbonate membranes in larger pore sizes are adaptable to many uses in cell culture including transport studies, toxicity tests, chemotaxis studies and electron microscopy. Membranes are transparent when wet and are suitable for phase contrast and Normanski Optic Systems.
  • Treated and quality controlled for cell culture
  • Excellent cell attachment and growth
  • Non toxic and resistant to most solvents
  • Low non specific binding
All Nunc™ Cell Culture Inserts: Recommended working volume (ml) = in addition to normal well working volume.