Optical bottom plates, 96-well, Nunclon™Δ
Supplier: Thermo Fisher Scientific
165306 165305 164588 164590
732-2605EA 680 EUR
732-2605 732-2604 734-2088 734-2089
Optical bottom plates, 96-well, Nunclon™Δ
Microplates Cell Culture Plates
PS plate with either polymer or coverglass base, sterile, with lid. These plates, with white or black upper structure and clear base, are designed to provide optimum clarity for viewing well contents.
  • Flat bottom well geometry for plate reader access
  • Footprint compatible with standard equipment and automated systems
  • Surface treatment ensures optimal cell attachment and growth
  • No. 1.5 coverglass base for minimum light scatter and low autofluorescence ensures accurate results due to higher signal to noise ratios
Working range: 50 to 200 µl/well
Working range: 50 - 200 µl/well