Lids for microplates, Whatman™
Supplier: Whatman products (Cytiva)
Whatman™
512-1093EA 170 EUR
512-1093
Lids for microplates, Whatman™
Lids Microplate Lids
Universal lids suitable for robotic handling. Can be used as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory.
Universal lids suitable for robotic handling. Can be used as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory.