Nunc-Immuno™ modules
Supplier: Thermo Fisher Scientific
441653 444865 445101 467120 467140 467466 467679 469078 469264 469914 469922 469949 469957 473709 475078 475086 468667 434797
735-0015EA 542 EUR
735-0015 735-0020 735-0021 735-0042 735-0043 735-0126 735-0046 735-0049 735-0050 735-0052 735-0053 735-0054 735-0055 735-0063 735-0066 735-0067 735-0048 737-0240
Nunc-Immuno™ modules
Microplates Immunology Microplates and Strips
PS. Nunc-Immuno™ modules have been developed for ELISA techniques where the demands of versatility require a different solution to the standard 96-well plate format. Available with a choice of MaxiSorp™, PolySorp™, MediSorp™ and MultiSorp™ surfaces. All MaxiSorp™ modules are certified for binding homogeneity. Framed modules (external dimensions: 128×86 mm) fit all standard equipment.
  • With Nunc StarWell™ modules incubation time can be reduced by more than 50% or the sensitivity similarly increased
  • Small numbers of samples can be processed more economically
  • Available framed or unframed, modules remain in the frame when inverted
  • Choice of well shape: Round (U), flat (F) or C-shaped (flat with curved edges)
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains. PolySorp™ surface has a high affinity for hydrophobic molecules. MediSorp™ has a surface chemistry intermediate between PolySorp™ and MaxiSorp™, providing low background signal with samples containing serum. MultiSorp™ surfaces have a high affinity to hydrophilic molecules.
Colour coded modules available on request, please visit www.vwr.com or contact your local VWR sales office for further information.