Modules, Nunc-Immuno™ LockWell™
Supplier: Thermo Fisher Scientific
446442 446469 446470 446490 446639 448496 448526 477089
735-0023EA 642 EUR
735-0023 735-0024 735-0025 732-2685 735-0027 735-0028 735-0029 391-7755
Modules, Nunc-Immuno™ LockWell™
Microplates Immunology Microplates and Strips
PS. Nunc-Immuno™ LockWell™ modules facilitate assays from large scale screening to single-well tests. Available with a choice of MaxiSorp™, PolySorp™, MediSorp™ and MultiSorp™ surfaces. All MaxiSorp™ modules are certified for binding homogeneity. Framed modules (external dimensions: 128×86 mm) fit all standard equipment.
  • Each well locks in the frame at the same height to prevent jamming in automated systems
  • Strips are easy to take out or put in the frame, without use of any special tools
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains; PolySorp™ surface has a high affinity for hydrophobic molecules; MediSorp™ has a surface chemistry intermediate between PolySorp™ and MaxiSorp™, providing low background signal with samples containing serum; MultiSorp™ surfaces have a high affinity to hydrophilic molecules.
Colour coded modules available on request, please visit website or contact your local Avantor sales office for further information.