Immunoassay microplates
Supplier: Thermo Fisher Scientific
95029390 3355 3455 3555 3655 3855 6301 6302 6309 6310 6404 6405 9502227 9502887 95029100 95029140 95029180 95029330
735-0551EA 329 EUR
735-0551 735-0461 735-0462 735-0463 735-0464 735-0465 735-0491 735-0492 735-0493 735-0494 735-0499 735-0500 735-0531 735-0535 735-0537 735-0539 735-0540 735-0546
Immunoassay microplates
Microplates Immunology Microplates and Strips
Designed specifically for immunoassay applications, this range of products have binding surfaces that include Immulon®, universal binding (UB) and enhanced binding (EB). These make good experimental design easier, allowing development of assays with stringent pH, buffer, pKa and resin requirements.
  • Available in 96-well or strip plate format for matching to the requirements of automated instruments
  • Microplates with breakable strips offer maximum efficiency for processing both large and small sample numbers, as the exact number of wells required can be used
  • Break-apart wells have a unique well locking system that keeps the wells and samples securely in place
  • Strips available in 1×8 and 1×12 break-apart or solid strip format, with paired 2×8 strips also available
Immulon® 1 B and UB microplates have medium binding surfaces with a binding capacity of 100 to 200 ng IgG/cm². Both have an affinity for hydrophobic complexes and proteins. Molecules for medium binding surfaces include polyclonal antibodies, proteins, immunoglobulins, lipids, lipoproteins and lipopolysaccharides.
Immulon® 2 HB, Immulon® 4 HBX and EB microplates have high binding, irradiated surfaces, with a binding capacity of 400 to 500 ng IgG/cm². Immulon® 4 HBX microplates have an extra surface with special resin for maximum protein uptake. EB microplates provide guaranteed coefficient of variation. Molecules for high binding surfaces include monoclonal and polyclonal antibodies, proteins, peptides, polysaccharides, glycoproteins, immunoglobulins, lipoproteins, glycoproteins and purified antigens.