VWR®, Cleanroom Mops
Supplier: VWR Collection
129-0147EA 552 EUR
129-0147 129-0148 129-0149
VWR®, Cleanroom Mops
Mops
100% polyester string mops. Double clear poly bagged for cleanroom use. Good chemical sorbancy. Ideal for application of cleaning solutions on floors in critical environments.
  • The sterile tubular knit mop, 129-0148, has a sterility assurance level of 10⁻⁶
  • The ribbed tubular knit mop offers the best sorbancy with chemical
Suitable for use in Class 10, ISO 4, M2.5 environments or below.