Sealing rings for conical ground glass joints
Supplier: GLINDEMANN
K5 K7 K10 K12 K14 K19 K24 K29 K34 K45 K60 K70
201-6250EA 56.1 EUR
201-6250 201-6251 201-6252 201-6253 201-6256 201-6258 201-6260 201-6262 201-6263 201-6264 201-6265 201-6266
Sealing rings for conical ground glass joints
Seals Sealing Rings
PTFE, white.
  • High-vacuum and chemical tight seals, without grease
  • Can be used at temperatures up to 300 °C
  • Can be combined with all joint clamps
These elasticated sealing rings adhere to the conical ground joint without a groove, and seal the joint, in a similar way to a screw cap.