Graduated pipettes, type 3, Class B, total delivery
Supplier: HIRSCHMANN
1030133 1030141
612-1022EA 7.7 EUR
612-1022 612-1024
Graduated pipettes, type 3, Class B, total delivery
Pipettes Graduated Pipettes
AR-Glas®
  • Calibrated to deliver 'Ex', short line graduation
  • Marks and inscriptions in amber stain, zero at the top, graduated to the tip
  • With cotton plug end
DIN EN ISO 835
Length: 360 mm ±5 mm
Certifikace: DIN EN ISO 835