Pipette controller, Pipette Pump™ II, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F379111002 F379111010 F379111025
612-0015EA 68.8 EUR
612-0015 612-0016 612-0017
Pipette controller, Pipette Pump™ II, SP Bel-Art
Pipettes Pipette Controllers
Pipette filler for glass and plastic pipettes up to 2, 10 or 25 ml.
  • Pipettes are easily inserted into a unique collar with stabilising fingers to hold pipette securely
  • Easily disassembled for cleaning
  • Colour-coded
Designed for fast and efficient single-handed operation with glass or plastic pipettes.