Pipette controller, pipetus®
Supplier: Hirschmann Laborgeräte
pipetus®
612-1874EA 490 EUR
612-1874
Pipette controller, pipetus®
Pipettes Pipette Controllers
Pipette controller for glass and plastic pipettes from 0,1 to 200 ml.
  • Unique and ecologically sound with up to 50 hours of independent running, cadmium-free NiMH batteries
  • Coloured display allows visual check of the battery charging status, the selected pipetting mode (Ex or blow-out), and the selected speed
  • Safe thanks to the safety return valve and highly sensitive valves for precise meniscus adjustment
  • Silent due to vibrationless pump motor
  • Silicone adapter for entirely leakproof positioning of each pipette
This pipette has a hydrophobic PTFE membrane filter with a porosity of 0,2 µm, intelligent charging technology, no memory effect, and protection against overcharging the batteries ensured by microprocessor control. The 'Speed' key allows you to choose the ideal pipetting speed from the five speeds offered, with colour indicator.
Informace pro objednávání: Supplied complete with charger and plate, two PTFE filters, one safety valve and one wall bracket.