Pipette controller, micro
Supplier: Brand
612-1910EA 49.8 EUR
612-1910
Pipette controller, micro
Pipettes Pipette Controllers
Pipette filler for disposable micro pipettes with ring marks and small volume pipettes up to 1 ml, Ø at end <5 mm.
  • Micro controller reduces the risk of contamination and is autoclavable at +121 °C (20 minutes)
  • The built-in ejector permits allows ejection of contaminated disposable pipettes up to 50 µl
  • Lightweight, fits snugly in the hand