Pipette aid for micro pipettes
Supplier: Brand
612-1449EA 2.7 EUR
612-1449
Pipette aid for micro pipettes
Pipettes Pipette Bulbs
PET plastic tube, silicone adapter, and TPE suction bellows with venting hole.
  • Especially for BLAUBRAND® intraEND micro pipettes (>1 µl)