Pipette jar
Supplier: Kartell
612-1820EA 87.8 EUR
612-1820
Pipette jar
Jars Pipette Jars
PP, with PE lid.
  • Can be used without the lid to autoclave pipettes (20 minutes at 120 °C)
  • Tight-fitting lid for protecting pipettes from dust and during transport
  • Can easily be cut down in height or used as a container for soaking contaminated pipettes
Ø×H ext/int: 82×426/410 mm