VWR® Pipette Stands
Supplier: VWR Collection
631-0723EA 82.4 EUR
631-0723 631-0724 631-0725
VWR® Pipette Stands
Pipette Stands
Pipette stands made of transparent acrylic.
  • Three, four or six-place racks, accommodate most micropipettes
  • Width of upper notches: 25 mm
  • Width of lower notches: 20 mm