VWR®, Sterilisation Boxes for Pipettes
Supplier: VWR
VWR®
612-2069EA 133 EUR
612-2069 612-2070
VWR®, Sterilisation Boxes for Pipettes
Boxes Pipette Sterilisation Containers
Stainless steel, with round base, autoclavable.
  • Can be autoclaved at 121 °C and sterilised at 180 °C
  • Bottom and underside of the lid are coated in silicone to minimise the risk of damaging the pipettes during transport and sterilisation