VWR®, Pipette Tips, Universal
Supplier: VWR Collection
612-5755EA 18.4 EUR
612-5755 612-6786
VWR®, Pipette Tips, Universal
Pipette Tips
PP, autoclavable.
  • Manufactured without lubricants
  • Cadmium-free pigments
  • Environmentally friendly packaging system
Colour: Yellow (2 - 200 µl), blue (50 - 1000 µl)
Length: 50 mm (2 - 200 µl), 70 mm (50 - 1000 µl)