Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
022.7142 022.7718 022.7725 022.7726 022.7727 022.7729 022.7730 022.7731 022.7732 022.7733 022.7734
552-0061EA 62.1 EUR
552-0061 552-0039 552-0040 552-0041 552-0042 552-0043 551-7730 551-7731 551-7732 551-7733 551-7734
Glass capillaries and graduated micropipettes, disposable
Pipettes Microcapillaries
Disposable glass capillaries for manual sample application of 0,5, 1, 2, 5 or 10 µl. They can be handheld and positioned with the Multipurpose Spotting Guide (551-4230). They can also be used with Universal Capillary Holder (552-0044). Graduated 5 µl glass micropipettes are suitable for qualitative analysis on conventional TLC layers.
  • Colour coded vials
  • Convenient and user friendly
  • Micropipettes are graduated in microlitres