Rack for cryogenic vials, Nunc™
Supplier: Thermo Fisher Scientific
Nunc™
391-8531EA 538 EUR
391-8531
Rack for cryogenic vials, Nunc™
Racks Cryogenic Vial Racks
Polyphenyleneoxide, autoclavable.
Allows one handed operation when used with starfoot vials.