Test tube racks, Unwire™, Nalgene®
Supplier: Thermo Fisher Scientific
5976-0013 5976-0016 5976-0313 5976-0316 5976-0413 5976-0416 5976-0513 5976-0516
212-8161EA 24.4 EUR
212-8161 212-8171 212-8162 212-0046 212-8163 212-8173 212-8164 212-8174
Test tube racks, Unwire™, Nalgene®
Racks Tube Racks
PP, coloured.
  • Racks can be stacked when empty
  • Very good chemical resistance
  • Not autoclavable
These racks hold a full load of tubes securely in position. They do not float in the water bath.